Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@7629baf6
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
Gene Symbol:
ECHDC2
Gene Name:
enoyl-CoA hydratase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 52895910 - 52927212 on Build GRCh38
UniGene ID:
Hs.476319
Species:
Human
siRNA ID
HSS124259
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ECHDC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198961.1
NP_001185890.1
3
347
NM_001198962.1
NP_001185891.1
3
347
NM_001319958.1
NP_001306887.1
4
860
NM_018281.3
NP_060751.2
3
347
XM_011541709.2
XP_011540011.1
4
558
XM_011541710.2
XP_011540012.1
4
558
XM_011541711.2
XP_011540013.1
4
558
XM_011541712.2
XP_011540014.1
4
558
XM_011541713.2
XP_011540015.1
3
4335
XM_011541715.2
XP_011540017.1
4
783
XM_011541719.2
XP_011540021.1
4
1780
XM_011541720.2
XP_011540022.1
3
4104
XM_011541721.2
XP_011540023.1
5
797
XM_011541722.2
XP_011540024.1
4
415
XM_011541723.2
XP_011540025.1
4
515
XM_011541724.2
XP_011540026.1
4
558
XM_011541725.2
XP_011540027.1
4
558
XM_011541726.2
XP_011540028.1
3
4335
XM_011541727.2
XP_011540029.1
3
4104
XM_017001638.1
XP_016857127.1
5
2169
XM_017001639.1
XP_016857128.1
4
574
XM_017001640.1
XP_016857129.1
3
737
XM_017001641.1
XP_016857130.1
5
1938
XM_017001642.1
XP_016857131.1
3
4335
GenBank mRNA
AK001810.1
-
3
299
AK299834.1
-
4
516
AK300896.1
-
4
386
AK302285.1
-
6
926
AK307381.1
-
3
242
AK310650.1
-
4
479
AK315906.1
-
4
478
BC038389.1
-
4
778
BC044574.1
-
3
277
5314083
BC051887.1
-
3
335
CF127686.1
-
3
291
Back To Top
More Information
Panther Classification:
Molecular Function -
acetyltransferase
acyltransferase
dehydrogenase
epimerase/racemase
hydratase
isomerase
ligase
lyase
oxidoreductase
transferase
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-2-prod-green:8080/10.253.226.220:8080.
git-commit: 0914912cb48844dc9118fffd0bcf8fc2c0fe1c71
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.17.0-2021.02.06-1.0