Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP6R3 SIRNA Assays ›
          Gene Symbol:
          PPP6R3
          Gene Name:
          protein phosphatase 6 regulatory subunit 3
          Gene Aliases:
          C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
          Chromosome Location:
          Chr.11: 68460712 - 68615334 on Build GRCh38
          UniGene ID:
          Hs.503022
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS124312
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP6R3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164160.1 NP_001157632.1 25 2881
          NM_001164161.1 NP_001157633.1 24 2863
          NM_001164162.1 NP_001157634.1 24 2845
          NM_001164163.1 NP_001157635.1 23 2776
          NM_018312.4 NP_060782.2 23 2728
          XM_006718608.3 XP_006718671.1 25 3009
          XM_006718612.3 XP_006718675.1 25 2991
          XM_006718613.2 XP_006718676.1 25 2979
          XM_006718614.2 XP_006718677.1 25 2961
          XM_006718618.3 XP_006718681.1 24 2892
          XM_006718621.2 XP_006718684.1 24 2841
          XM_006718622.3 XP_006718685.1 23 2769
          XM_006718623.3 XP_006718686.1 23 2752
          XM_006718624.2 XP_006718687.1 23 2739
          XM_011545135.1 XP_011543437.1 26 3027
          XM_011545136.2 XP_011543438.1 25 2910
          XM_011545137.2 XP_011543439.1 26 3010
          XM_011545138.2 XP_011543440.1 27 3228
          XM_011545139.1 XP_011543441.1 26 3009
          XM_011545140.2 XP_011543442.1 26 2997
          XM_011545141.1 XP_011543443.1 26 2976
          XM_011545142.2 XP_011543444.1 25 2940
          XM_011545143.1 XP_011543445.1 25 2922
          XM_011545145.2 XP_011543447.1 25 2874
          XM_017017962.1 XP_016873451.1 25 2835
          XM_017017963.1 XP_016873452.1 25 2903
          XM_017017964.1 XP_016873453.1 26 2979
          XM_017017965.1 XP_016873454.1 25 2862
          XM_017017966.1 XP_016873455.1 25 3047
          XM_017017967.1 XP_016873456.1 26 2946
          XM_017017968.1 XP_016873457.1 25 2829
          XM_017017969.1 XP_016873458.1 25 2928
          XM_017017970.1 XP_016873459.1 24 2811
          XM_017017971.1 XP_016873460.1 25 2910
          XM_017017972.1 XP_016873461.1 24 2766
          XM_017017973.1 XP_016873462.1 25 2893
          XM_017017974.1 XP_016873463.1 25 2892
          XM_017017975.1 XP_016873464.1 25 2859
          XM_017017976.1 XP_016873465.1 23 2658
          XM_017017977.1 XP_016873466.1 25 2768
          XM_017017978.1 XP_016873467.1 24 2619
          XM_017017979.1 XP_016873468.1 23 2613
          XM_017017980.1 XP_016873469.1 23 2601
          XM_017017981.1 XP_016873470.1 23 2550
          XM_017017982.1 XP_016873471.1 22 2514
          XM_017017983.1 XP_016873472.1 24 3000
          XM_017017984.1 XP_016873473.1 23 2843
          GenBank mRNA AB046778.2 - 25 2829
          AF264779.1 - 22 2633
          AK001920.1 - 5 542
          AK023950.1 - 7 769
          AK057250.1 - 18 1936
          AK125055.1 - 10 2633
          AK300359.1 - 16 1697
          AK302611.1 - 16 1717
          AK303315.1 - 19 2179
          AK315136.1 - 23 2745
          AL389982.1 - 1 1472
          AL834471.1 - 7 740
          BC007738.2 - 6 651
          BC020953.1 - 10 985
          BC105933.1 - 23 2731 40029064
          BC105934.1 - 23 2731 40029058
          BC107599.1 - 22 2644 40029065
          CR749815.1 - 24 2808
          CR933658.1 - 24 2826
          CR936718.1 - 24 2831
          DQ111954.2 - 22 2596

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of phosphoprotein phosphatase activity
          COPII vesicle coating
          protein binding
          protein phosphatase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0