Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP6R3 SIRNA Assays ›
Gene Symbol:
PPP6R3
Gene Name:
protein phosphatase 6 regulatory subunit 3
Gene Aliases:
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
Chromosome Location:
Chr.11: 68460712 - 68615334 on Build GRCh38
UniGene ID:
Hs.503022
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS124314
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP6R3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164160.1
NP_001157632.1
4, 5
683
NM_001164161.1
NP_001157633.1
4, 5
683
NM_001164162.1
NP_001157634.1
4, 5
683
NM_001164163.1
NP_001157635.1
4, 5
683
NM_001164164.1
NP_001157636.1
5, 6
788
NM_018312.4
NP_060782.2
5, 6
788
XM_006718608.3
XP_006718671.1
5, 6
799
XM_006718612.3
XP_006718675.1
5, 6
799
XM_006718613.2
XP_006718676.1
5, 6
799
XM_006718614.2
XP_006718677.1
5, 6
799
XM_006718618.3
XP_006718681.1
5, 6
799
XM_006718621.2
XP_006718684.1
5, 6
799
XM_006718622.3
XP_006718685.1
5, 6
799
XM_006718623.3
XP_006718686.1
5, 6
782
XM_006718624.2
XP_006718687.1
5, 6
799
XM_006718627.3
XP_006718690.1
5, 6
799
XM_011545135.1
XP_011543437.1
5, 6
799
XM_011545136.2
XP_011543438.1
4, 5
682
XM_011545137.2
XP_011543439.1
5, 6
782
XM_011545138.2
XP_011543440.1
6, 7
1000
XM_011545139.1
XP_011543441.1
5, 6
799
XM_011545140.2
XP_011543442.1
5, 6
799
XM_011545141.1
XP_011543443.1
5, 6
799
XM_011545142.2
XP_011543444.1
5, 6
799
XM_011545143.1
XP_011543445.1
5, 6
799
XM_011545145.2
XP_011543447.1
5, 6
799
XM_017017962.1
XP_016873451.1
4, 5
637
XM_017017963.1
XP_016873452.1
5, 6
723
XM_017017964.1
XP_016873453.1
5, 6
799
XM_017017965.1
XP_016873454.1
4, 5
682
XM_017017966.1
XP_016873455.1
5, 6
885
XM_017017967.1
XP_016873456.1
5, 6
799
XM_017017968.1
XP_016873457.1
4, 5
682
XM_017017969.1
XP_016873458.1
5, 6
799
XM_017017970.1
XP_016873459.1
4, 5
682
XM_017017971.1
XP_016873460.1
5, 6
799
XM_017017972.1
XP_016873461.1
4, 5
655
XM_017017973.1
XP_016873462.1
5, 6
782
XM_017017974.1
XP_016873463.1
5, 6
799
XM_017017975.1
XP_016873464.1
5, 6
799
XM_017017976.1
XP_016873465.1
4, 5
682
XM_017017977.1
XP_016873466.1
4, 5
570
XM_017017978.1
XP_016873467.1
3, 4
421
XM_017017979.1
XP_016873468.1
3, 4
421
XM_017017980.1
XP_016873469.1
3, 4
421
XM_017017981.1
XP_016873470.1
3, 4
421
XM_017017982.1
XP_016873471.1
3, 4
421
XM_017017983.1
XP_016873472.1
5, 6
802
XM_017017984.1
XP_016873473.1
4, 5
619
GenBank mRNA
AB046778.2
-
4, 5
631
AF264779.1
-
4, 5
693
AF264780.1
-
4, 5
693
AK298180.1
-
4, 5
548
AK303315.1
-
3, 4
422
AK315136.1
-
4, 5
652
BC105933.1
-
3, 4
551
40029064
BC105934.1
-
3, 4
551
40029058
BC107599.1
-
3, 4
551
40029065
CR749815.1
-
4, 5
646
CR933658.1
-
4, 5
646
CR936718.1
-
4, 5
651
DQ111954.2
-
2, 3
416
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of phosphoprotein phosphatase activity
COPII vesicle coating
protein binding
protein phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0