Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018711.4 | NP_061181.1 | 13 | 1614 | ||
GenBank mRNA | AK309780.1 | - | 4 | 359 | ||
AL359592.1 | - | 1 | 32 | |||
BC033587.1 | - | 13 | 1414 | 5243017 | ||
BC094722.1 | - | 13 | 1261 | 4817829 |