Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018725.3 | NP_061195.2 | 10, 11 | 993 | ||
XM_005265310.4 | XP_005265367.1 | 11, 12 | 1080 | |||
XM_005265311.4 | XP_005265368.1 | 10, 11 | 1057 | |||
XM_005265312.4 | XP_005265369.1 | 9, 10 | 1001 | |||
XM_017006804.1 | XP_016862293.1 | 10, 11 | 949 | |||
XM_017006805.1 | XP_016862294.1 | 9, 10 | 901 | |||
XM_017006806.1 | XP_016862295.1 | 9, 10 | 862 | |||
GenBank mRNA | AF208110.1 | - | 10, 11 | 959 | ||
AF208111.1 | - | 9, 10 | 1988 | |||
AF212365.1 | - | 10, 11 | 963 | |||
AF250309.1 | - | 10, 11 | 954 | |||
AK307749.1 | - | 9, 10 | 951 | |||
AY359122.1 | - | 10, 11 | 960 | |||
BC000980.1 | - | 10, 11 | 963 | 2985728 |