Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SUPT20H SIRNA Assays ›
Gene Symbol:
SUPT20H
Gene Name:
SPT20 homolog, SAGA complex component
Gene Aliases:
C13, C13orf19, FAM48A, FP757, P38IP, SPT20
Chromosome Location:
Chr.13: 37009312 - 37059714 on Build GRCh38
UniGene ID:
Hs.435815
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS124608
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SUPT20H
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014286.2
NP_001014308.2
23
2181
NM_001278480.1
NP_001265409.1
23
2418
NM_001278481.1
NP_001265410.1
23
2184
NM_001278482.1
NP_001265411.1
23
2322
NM_017569.3
NP_060039.1
23
2263
XM_005266447.2
XP_005266504.1
23
2394
XM_005266449.3
XP_005266506.1
23
2355
XM_005266451.3
XP_005266508.1
23
2391
XM_005266454.3
XP_005266511.1
22
2353
XM_005266455.2
XP_005266512.1
23
2358
XM_005266456.3
XP_005266513.1
22
2426
XM_005266457.3
XP_005266514.1
22
2352
XM_005266458.3
XP_005266515.1
22
2423
XM_005266460.2
XP_005266517.1
23
2394
XM_005266461.3
XP_005266518.1
23
2358
XM_005266462.4
XP_005266519.1
23
2157
XM_005266464.3
XP_005266521.1
23
2121
XM_005266465.2
XP_005266522.1
23
2154
XM_005266467.3
XP_005266524.1
23
2121
XM_017020653.1
XP_016876142.1
23
2355
XM_017020654.1
XP_016876143.1
22
2116
XM_017020655.1
XP_016876144.1
22
2115
XM_017020656.1
XP_016876145.1
22
2184
XM_017020657.1
XP_016876146.1
23
2118
XM_017020658.1
XP_016876147.1
22
2136
XM_017020659.1
XP_016876148.1
22
2115
XM_017020660.1
XP_016876149.1
23
2118
XM_017020661.1
XP_016876150.1
22
2079
GenBank mRNA
AF093250.1
-
23
2129
AF370384.1
-
6
1110
AJ130894.1
-
23
2219
AK026091.1
-
8
728
AK056121.1
-
4
1231
AK225376.1
-
23
2354
AK292376.1
-
23
2267
BC001145.1
-
24
2307
3533877
BC030686.2
-
23
2156
4826077
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
autophagy
gastrulation
positive regulation of gluconeogenesis by positive regulation of transcription from RNA polymerase II promoter
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0