Search
Search

PACS1| Gene Symbol: PACS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018026.3 | NP_060496.2 | 21 | 2446 | ||
| XM_011545162.1 | XP_011543464.1 | 21 | 2006 | |||
| XM_011545164.2 | XP_011543466.1 | 21 | 3314 | |||
| GenBank mRNA | AB033001.1 | - | 14 | 1348 | ||
| AK001071.1 | - | 9 | 751 | |||
| AK056361.1 | - | 20 | 1953 | |||
| AK293966.1 | - | 10 | 1057 | |||
| AY320283.1 | - | 21 | 2486 | |||
| BC003173.1 | - | 6 | 415 | |||
| BC009936.1 | - | 7 | 521 | |||
| BC010096.1 | - | 21 | 2354 | 3502170 | ||
| BC042909.1 | - | 10 | 905 | |||
| BC052577.1 | - | 21 | 2345 | 4130626 | ||
| BC055288.1 | - | 13 | 1150 | |||