Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018128.4 | NP_060598.3 | 5 | 1656 | ||
GenBank mRNA | AB037822.1 | - | 5 | 1779 | ||
AK001396.1 | - | 3 | 298 | |||
AK026565.1 | - | 5 | 729 | |||
BC019090.2 | - | 4 | 564 | |||
BC110851.1 | - | 5 | 1234 | 5786818 | ||
BC126110.1 | - | 5 | 1204 | 8991826 |