Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASUN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018164.2 | NP_060634.2 | 10 | 1581 | ||
XM_005253422.3 | XP_005253479.1 | 10 | 1540 | |||
XM_011520750.2 | XP_011519052.1 | 10 | 1132 | |||
XM_011520751.2 | XP_011519053.1 | 10 | 1558 | |||
XM_011520752.2 | XP_011519054.1 | 10 | 1558 | |||
XM_011520753.2 | XP_011519055.1 | 4 | 304 | |||
XM_017019632.1 | XP_016875121.1 | 10 | 1114 | |||
XM_017019633.1 | XP_016875122.1 | 9 | 1524 | |||
XM_017019634.1 | XP_016875123.1 | 9 | 1506 | |||
XM_017019635.1 | XP_016875124.1 | 10 | 1114 | |||
XM_017019636.1 | XP_016875125.1 | 10 | 1540 | |||
XM_017019637.1 | XP_016875126.1 | 8 | 744 | |||
GenBank mRNA | AF274950.1 | - | 2 | 163 | ||
AK001222.1 | - | 1 | 66 | |||
AK001492.1 | - | 10 | 1210 | |||
AK001499.1 | - | 10 | 1246 | |||
AK225336.1 | - | 10 | 1232 | |||
AK297948.1 | - | 8 | 921 | |||
AY211927.1 | - | 3 | 241 | |||
BC003081.2 | - | 10 | 1270 | 2989678 | ||
BC008368.1 | - | 10 | 1228 | 3355282 |