Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM184C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018241.2 | NP_060711.2 | 9 | 1573 | ||
GenBank mRNA | AF305823.1 | - | 2 | 249 | ||
AK000169.1 | - | 6 | 726 | |||
AK001708.1 | - | 9 | 1516 | |||
BC008120.1 | - | 1 | 105 | |||
BM712423.1 | - | 3 | 293 |