Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBNL3 SIRNA Assays ›
Gene Symbol:
MBNL3
Gene Name:
muscleblind like splicing regulator 3
Gene Aliases:
CHCR, MBLX, MBLX39, MBXL
Chromosome Location:
Chr.X: 132369314 - 132490030 on Build GRCh38
UniGene ID:
Hs.105134
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS125060
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBNL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170701.1
NP_001164172.1
4
498
NM_001170702.1
NP_001164173.1
4
498
NM_001170703.1
NP_001164174.1
4
431
NM_001170704.1
NP_001164175.1
5
698
NM_018388.3
NP_060858.2
4
661
NM_133486.2
NP_597846.1
4
661
XM_005262431.3
XP_005262488.1
5
1473
XM_005262434.3
XP_005262491.1
5
1468
XM_005262436.3
XP_005262493.1
4
1361
XM_005262437.3
XP_005262494.1
4
499
XM_005262438.3
XP_005262495.1
4
499
XM_005262439.3
XP_005262496.1
4
431
XM_005262440.3
XP_005262497.1
6
1518
XM_005262441.3
XP_005262498.1
4
622
XM_005262442.3
XP_005262499.1
4
593
XM_011531364.2
XP_011529666.1
3
1509
XM_017029632.1
XP_016885121.1
5
8903
XM_017029633.1
XP_016885122.1
6
1638
XM_017029634.1
XP_016885123.1
5
8903
XM_017029635.1
XP_016885124.1
5
8903
XM_017029636.1
XP_016885125.1
5
1402
XM_017029637.1
XP_016885126.1
5
8903
XM_017029638.1
XP_016885127.1
6
1536
XM_017029639.1
XP_016885128.1
4
611
GenBank mRNA
AB077698.1
-
4
661
AB077699.1
-
4
661
AF467070.1
-
4
661
AF491305.1
-
4
582
AJ427918.1
-
4
661
AJ427919.1
-
4
661
AK002178.1
-
4
467
AK300248.1
-
4
430
AL133625.1
-
3
406
AY072692.1
-
4
661
AY372211.1
-
4
432
BC042090.1
-
5
697
5761647
BC074775.2
-
4
624
30915333
BC074776.2
-
4
624
30915555
BU569439.1
-
4
501
DB023330.1
-
4
431
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
multicellular organism development
RNA splicing
regulation of RNA splicing
negative regulation of myoblast differentiation
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0