Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERBIN SIRNA Assays ›
          Gene Symbol:
          ERBIN
          Gene Name:
          erbb2 interacting protein
          Gene Aliases:
          ERBB2IP, HEL-S-78, LAP2
          Chromosome Location:
          Chr.5: 65926554 - 66081024 on Build GRCh38
          UniGene ID:
          Hs.591774
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS125240
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ERBIN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006600.2 NP_001006600.1 21 3067
          NM_001253697.1 NP_001240626.1 21 3067
          NM_001253698.1 NP_001240627.1 21 3067
          NM_001253699.1 NP_001240628.1 21 3067
          NM_001253701.1 NP_001240630.1 21 3055
          NM_018695.3 NP_061165.1 21 3067
          XM_005248554.2 XP_005248611.1 21 3022
          XM_005248555.3 XP_005248612.1 21 3010
          XM_006714660.3 XP_006714723.1 21 3022
          XM_017009635.1 XP_016865124.1 21 2838
          XM_017009636.1 XP_016865125.1 21 3010
          XM_017009637.1 XP_016865126.1 21 3010
          GenBank mRNA AB033051.2 - 19 2757
          AF263744.1 - 21 3080
          AF276423.1 - 21 2900
          AK001180.1 - 3 959
          AK304693.1 - 21 3013
          BC115012.1 - 20 2837 7961934
          BC115015.1 - 20 2825 7961940
          BC126464.1 - 21 2848 8992180
          BC144075.1 - 21 2848 9052595

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting
          cell cycle
          cell adhesion
          signal transduction
          epidermal growth factor receptor signaling pathway
          integrin-mediated signaling pathway
          cell growth
          negative regulation of NF-kappaB transcription factor activity
          response to muramyl dipeptide
          response to lipopolysaccharide
          ERBB2 signaling pathway
          intermediate filament cytoskeleton organization
          basal protein localization
          establishment or maintenance of epithelial cell apical/basal polarity
          negative regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
          cellular response to tumor necrosis factor
          negative regulation of monocyte chemotactic protein-1 production
          receptor binding
          ErbB-2 class receptor binding
          integrin binding
          structural constituent of cytoskeleton
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0