Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019117.4 | NP_061990.2 | 7 | 1544 | ||
NM_057162.2 | NP_476503.1 | 7 | 1544 | |||
GenBank mRNA | AB051474.1 | - | 7 | 1490 | ||
AF284765.1 | - | 7 | 1451 | |||
AF284766.1 | - | 7 | 1451 | |||
AL831868.1 | - | 2 | 203 | |||
BC161675.1 | - | 7 | 1490 | 8862604 |