Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC2A9 SIRNA Assays ›
Gene Symbol:
SLC2A9
Gene Name:
solute carrier family 2 member 9
Gene Aliases:
GLUT9, GLUTX, UAQTL2, URATv1
Chromosome Location:
Chr.4: 9771125 - 10040248 on Build GRCh38
UniGene ID:
Hs.444612
Species:
Human
siRNA ID
HSS125577
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC2A9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001290.1
NP_001001290.1
6, 7
763
NM_020041.2
NP_064425.2
5, 6
752
XM_006713968.3
XP_006714031.1
5, 6
864
XM_006713969.2
XP_006714032.1
6, 7
725
XM_011513856.2
XP_011512158.1
5, 6
889
XM_011513857.1
XP_011512159.1
6, 7
725
XM_011513858.1
XP_011512160.1
6, 7
927
XM_011513859.2
XP_011512161.1
5, 6
859
XM_011513860.2
XP_011512162.1
5, 6
859
XM_011513861.2
XP_011512163.1
5, 6
861
XM_011513862.2
XP_011512164.1
4, 5
736
XM_011513863.2
XP_011512165.1
4, 5
2415
XM_011513864.2
XP_011512166.1
3, 4
304
XM_011513865.2
XP_011512167.1
5, 6
863
XM_011513866.2
XP_011512168.1
5, 6
861
XM_011513867.2
XP_011512169.1
2, 3
818
XM_011513868.2
XP_011512170.1
5, 6
880
XM_017008457.1
XP_016863946.1
5, 6
877
XM_017008458.1
XP_016863947.1
5, 6
862
XM_017008459.1
XP_016863948.1
3, 4
396
XM_017008460.1
XP_016863949.1
4, 5
2409
GenBank mRNA
AF210317.1
-
5, 6
745
AF421859.1
-
3, 4
440
AK310895.1
-
6, 7
725
BC018897.2
-
6, 7
763
3949549
BC110414.1
-
5, 6
782
5762360
CD629822.1
-
1
106
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
glucose transport
proton transport
urate metabolic process
organic acid transmembrane transport
glucose transmembrane transport
sugar:proton symporter activity
glucose transmembrane transporter activity
urate transmembrane transporter activity
transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0