Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020129.2 | NP_064514.1 | 4 | 556 | ||
NM_203471.1 | NP_982297.1 | 5 | 861 | |||
GenBank mRNA | AF123762.1 | - | 3 | 333 | ||
AF267852.1 | - | 4 | 556 | |||
AI167958.1 | - | 1 | 218 | |||
AK312040.1 | - | 4 | 395 | |||
BC022257.1 | - | 4 | 412 | 4770140 | ||
BQ007347.1 | - | 1 | 235 | |||
FJ613341.1 | - | 4 | 360 | |||
FJ613342.1 | - | 4 | 360 |