Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015492.4 | NP_056307.2 | 2 | 2304 | ||
GenBank mRNA | AF495726.1 | - | 2 | 2289 | ||
AK056589.1 | - | 1 | 818 | |||
AK057025.1 | - | 1 | 179 | |||
AK123746.1 | - | 1 | 1679 | |||
AK125111.1 | - | 2 | 2302 | |||
AK125604.1 | - | 2 | 2273 | |||
AK128205.1 | - | 2 | 2394 | |||
AK304663.1 | - | 2 | 755 | |||
AL360143.1 | - | 1 | 536 | |||
AL834382.1 | - | 1 | 396 | |||
BC000540.2 | - | 1 | 442 | |||
BC006207.2 | - | 1 | 1004 | |||
BC011379.1 | - | 2 | 2276 | 3866082 | ||
BC109195.1 | - | 1 | 762 | |||
BC110367.1 | - | 2 | 2291 | 5494248 |