Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020188.3 | NP_064573.1 | 4 | 392 | ||
XM_017023461.1 | XP_016878950.1 | 5 | 580 | |||
XM_017023462.1 | XP_016878951.1 | 6 | 705 | |||
XM_017023464.1 | XP_016878953.1 | 3 | 449 | |||
XM_017023465.1 | XP_016878954.1 | 4 | 568 | |||
XM_017023466.1 | XP_016878955.1 | 2 | 353 | |||
XM_017023467.1 | XP_016878956.1 | 6 | 475 | |||
XM_017023468.1 | XP_016878957.1 | 5 | 385 | |||
XM_017023469.1 | XP_016878958.1 | 5 | 459 | |||
XM_017023470.1 | XP_016878959.1 | 5 | 383 | |||
GenBank mRNA | AF201935.1 | - | 4 | 368 | ||
AK123193.1 | - | 3 | 1747 | |||
BC032631.1 | - | 4 | 396 | 5399193 |