Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020247.4 | NP_064632.2 | 2 | 321 | ||
XM_005273201.1 | XP_005273258.1 | 2 | 217 | |||
XM_011544238.1 | XP_011542540.1 | 3 | 413 | |||
XM_011544239.2 | XP_011542541.1 | 4 | 436 | |||
XM_011544240.2 | XP_011542542.1 | 2 | 230 | |||
XM_011544241.2 | XP_011542543.1 | 2 | 192 | |||
XM_017001852.1 | XP_016857341.1 | 2 | 411 | |||
GenBank mRNA | AB073905.1 | - | 2 | 244 | ||
AK074693.1 | - | 2 | 264 | |||
AK293571.1 | - | 2 | 264 | |||
AK301012.1 | - | 3 | 278 | |||
BC005171.2 | - | 2 | 270 | 3604948 | ||
BC129930.1 | - | 8 | 1163 | 40010265 | ||
BX648860.1 | - | 2 | 142 | |||
DA925261.1 | - | 2 | 321 |