Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RTN4 SIRNA Assays ›
Gene Symbol:
RTN4
Gene Name:
reticulon 4
Gene Aliases:
ASY, NI220/250, NOGO, NOGO-A, NOGOC, NSP, NSP-CL, Nbla00271, Nbla10545, Nogo-B, Nogo-C, RTN-X, RTN4-A, RTN4-B1, RTN4-B2, RTN4-C
Chromosome Location:
Chr.2: 54972187 - 55137831 on Build GRCh38
UniGene ID:
Hs.637850
Species:
Human
siRNA ID
HSS126050
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RTN4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321859.1
NP_001308788.1
4
2729
NM_001321860.1
NP_001308789.1
4
2799
NM_001321861.1
NP_001308790.1
4
2762
NM_001321862.1
NP_001308791.1
4
2749
NM_001321863.1
NP_001308792.1
4
2699
NM_001321904.1
NP_001308833.1
5
3177
NM_007008.2
NP_008939.1
2
419
NM_020532.4
NP_065393.1
4
3482
NM_153828.2
NP_722550.1
2
1025
NM_207520.1
NP_997403.1
3
1082
NM_207521.1
NP_997404.1
4
2740
NR_135829.1
2
305
NR_135830.1
2
283
XM_005264434.3
XP_005264491.1
5
2876
XM_017004518.1
XP_016860007.1
5
2915
XM_017004519.1
XP_016860008.1
5
3032
GenBank mRNA
AB015639.1
-
2
793
AB020693.1
-
4
3318
AB040462.1
-
4
3428
AB040463.1
-
2
971
AB073351.1
-
2
1154
AF063601.1
-
2
1742
AF077050.1
-
2
418
AF087901.1
-
2
310
AF125103.1
-
2
419
AF132047.1
-
2
906
AF132048.1
-
2
325
AF148537.1
-
4
3325
AF148538.1
-
2
868
AF177332.1
-
2
324
AF320999.1
-
3
2488
AF333336.1
-
4
2744
AJ251383.1
-
4
3184
AJ251384.1
-
2
727
AJ251385.1
-
2
205
AK075039.1
-
2
959
AK129806.1
-
2
884
AK130812.1
-
2
786
AY102276.1
-
2
439
AY102277.1
-
2
971
AY102278.1
-
3
1028
AY102279.1
-
4
3428
AY123245.1
-
4
2741
AY123246.1
-
4
2799
AY123247.1
-
4
2762
AY123248.1
-
4
2748
AY123249.1
-
4
2709
AY123250.1
-
4
2699
BC001035.2
-
2
390
BC007109.1
-
2
316
BC010737.1
-
2
732
BC012619.1
-
2
882
BC014366.1
-
2
316
BC016165.1
-
3
1031
BC026788.1
-
2
913
BC068991.1
-
2
975
BC071848.1
-
3
713
BC150182.1
-
4
3432
40148210
BC152425.1
-
4
3318
8862539
BC152555.1
-
4
3432
40148984
CN429712.1
-
2
254
DQ778739.1
-
2
205
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
axonal fasciculation
cerebral cortex radial glia guided migration
negative regulation of cell growth
negative regulation of axon extension
regulation of apoptotic process
negative regulation of axonogenesis
nuclear pore complex assembly
cardiac epithelial to mesenchymal transition
endoplasmic reticulum tubular network organization
endoplasmic reticulum tubular network assembly
cell-cell adhesion
regulation of branching morphogenesis of a nerve
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0