Search
Search

TRIM54| Gene Symbol: TRIM54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032546.3 | NP_115935.3 | 3 | 707 | ||
| NM_187841.2 | NP_912730.2 | 3 | 707 | |||
| GenBank mRNA | AJ291714.1 | - | 3 | 404 | ||
| AK300699.1 | - | 3 | 708 | |||
| BC141807.1 | - | 3 | 479 | 40147443 | ||