Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021255.2 | NP_067078.1 | 1, 2 | 365 | ||
XM_005267890.4 | XP_005267947.1 | 1, 2 | 410 | |||
GenBank mRNA | AF302502.1 | - | 1, 2 | 255 | ||
AK315727.1 | - | 1, 2 | 327 | |||
BC009476.2 | - | 1, 2 | 365 | 3942712 |