Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032394.2 | NP_001027566.1 | 12 | 2129 | ||
NM_001032395.2 | NP_001027567.1 | 12 | 2129 | |||
NM_020455.5 | NP_065188.4 | 13 | 2213 | |||
NM_198569.2 | NP_940971.1 | 13 | 2213 | |||
XM_005267061.3 | XP_005267118.1 | 13 | 2114 | |||
XM_006715516.2 | XP_006715579.1 | 13 | 2114 | |||
XM_006715517.2 | XP_006715580.1 | 13 | 1914 | |||
XM_006715518.2 | XP_006715581.1 | 12 | 2030 | |||
XM_011535964.1 | XP_011534266.1 | 13 | 1917 | |||
XM_017011085.1 | XP_016866574.1 | 13 | 2114 | |||
GenBank mRNA | AB183546.1 | - | 13 | 2203 | ||
AB183547.1 | - | 12 | 2119 | |||
AB183548.1 | - | 13 | 2203 | |||
AB183549.1 | - | 12 | 2119 | |||
AK307543.1 | - | 12 | 2027 | |||
BC075798.1 | - | 12 | 2189 | 30333185 | ||
BX640873.1 | - | 12 | 1735 | |||
BX640971.1 | - | 13 | 2139 | |||
BX648315.1 | - | 12 | 1842 |
Set Membership: |
G-protein coupled receptor |