Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNX14 SIRNA Assays ›
Gene Symbol:
SNX14
Gene Name:
sorting nexin 14
Gene Aliases:
RGS-PX2, SCAR20
Chromosome Location:
Chr.6: 85505496 - 85594156 on Build GRCh38
UniGene ID:
Hs.485871
Species:
Human
siRNA ID
HSS126179
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNX14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001297614.2
NP_001284543.1
24
2834
NM_001304479.1
NP_001291408.1
25
2606
NM_020468.5
NP_065201.1
22
2702
NM_153816.5
NP_722523.1
25
2861
XM_005248738.3
XP_005248795.1
25
2834
XM_005248740.3
XP_005248797.1
23
2703
XM_011535977.2
XP_011534279.1
24
2277
XM_011535979.1
XP_011534281.1
23
2269
XM_017011090.1
XP_016866579.1
25
2343
XM_017011091.1
XP_016866580.1
24
2807
XM_017011092.1
XP_016866581.1
23
2471
XM_017011093.1
XP_016866582.1
25
2548
XM_017011094.1
XP_016866583.1
23
2443
XM_017011095.1
XP_016866584.1
23
2183
XM_017011096.1
XP_016866585.1
25
2572
XM_017011097.1
XP_016866586.1
25
2556
XM_017011098.1
XP_016866587.1
24
2474
XM_017011099.1
XP_016866588.1
23
2440
XM_017011100.1
XP_016866589.1
22
2164
XM_017011101.1
XP_016866590.1
23
2246
GenBank mRNA
AF121863.1
-
8
707
AJ420561.1
-
2
94
AK000362.1
-
25
2550
AK026479.1
-
23
2966
AK095380.1
-
25
3030
AK125063.1
-
21
2913
AK226097.1
-
13
1172
AK295417.1
-
25
2335
AY044865.1
-
22
2243
BC005110.1
-
22
2431
3959086
BC046520.2
-
24
2289
BC068589.1
-
25
2600
4818366
BC095419.1
-
25
2446
5268442
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein transport
autophagosome maturation
phosphatidylinositol binding
phosphatidylinositol-3,5-bisphosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-hf9rh:80/10.239.161.5:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0