Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDRG2 SIRNA Assays ›
Gene Symbol:
NDRG2
Gene Name:
NDRG family member 2
Gene Aliases:
SYLD
Chromosome Location:
Chr.14: 21016763 - 21070872 on Build GRCh38
UniGene ID:
Hs.525205
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS126268
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDRG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282211.1
NP_001269140.1
6, 7
468
NM_001282212.1
NP_001269141.1
7, 8
742
NM_001282213.1
NP_001269142.1
7, 8
747
NM_001282214.1
NP_001269143.1
7, 8
735
NM_001282215.1
NP_001269144.1
7, 8
565
NM_001282216.1
NP_001269145.1
6, 7
483
NM_001320329.1
NP_001307258.1
7, 8
1400
NM_016250.2
NP_057334.1
7, 8
590
NM_201535.1
NP_963293.1
8, 9
632
NM_201536.1
NP_963294.1
6, 7
523
NM_201537.1
NP_963831.1
7, 8
565
NM_201538.1
NP_963832.1
6, 7
500
NM_201539.1
NP_963833.1
7, 8
542
NM_201540.1
NP_963834.1
8, 9
609
NM_201541.1
NP_963835.1
7, 8
567
XM_011536996.2
XP_011535298.1
8, 9
619
XM_011536997.1
XP_011535299.1
8, 9
620
XM_011536998.1
XP_011535300.1
8, 9
610
XM_011536999.1
XP_011535301.1
6, 7
1015
XM_011537001.1
XP_011535303.1
7, 8
573
XM_011537002.1
XP_011535304.1
7, 8
719
XM_017021480.1
XP_016876969.1
8, 9
571
XM_017021481.1
XP_016876970.1
8, 9
610
XM_017021482.1
XP_016876971.1
8, 9
619
XM_017021483.1
XP_016876972.1
7, 8
503
XM_017021484.1
XP_016876973.1
7, 8
545
XM_017021485.1
XP_016876974.1
8, 9
610
XM_017021486.1
XP_016876975.1
8, 9
620
XM_017021487.1
XP_016876976.1
7, 8
529
XM_017021488.1
XP_016876977.1
7, 8
570
XM_017021489.1
XP_016876978.1
7, 8
675
XM_017021490.1
XP_016876979.1
6, 7
467
XM_017021491.1
XP_016876980.1
6, 7
502
XM_017021492.1
XP_016876981.1
7, 8
573
XM_017021493.1
XP_016876982.1
7, 8
719
XM_017021494.1
XP_016876983.1
8, 9
571
XM_017021495.1
XP_016876984.1
7, 8
503
XM_017021496.1
XP_016876985.1
6, 7
502
XM_017021497.1
XP_016876986.1
2, 3
100
GenBank mRNA
AB033074.1
-
6, 7
452
AF087872.1
-
6, 7
506
AF159092.3
-
7, 8
513
AF304051.1
-
8, 9
632
AK024521.1
-
8, 9
565
AK057843.1
-
7, 8
536
AK057847.1
-
6, 7
500
AK090657.1
-
6, 7
439
AK096999.1
-
7, 8
1400
AK130029.1
-
6, 7
438
AK293514.1
-
6, 7
468
AL136574.1
-
6, 7
477
BC010458.1
-
6, 7
471
4215141
BC011240.1
-
7, 8
525
4153602
BC093038.1
-
7, 8
566
4811718
BG818977.1
-
7, 8
546
BI759006.1
-
8, 9
551
BM810623.1
-
7, 8
506
BX247987.1
-
7, 8
601
BX647091.1
-
4, 5
665
BX647748.1
-
7, 8
682
CB153966.1
-
6, 7
566
CR749252.1
-
7, 8
735
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cytokine production
signal transduction
regulation of vascular endothelial growth factor production
Wnt signaling pathway
substantia nigra development
cell differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of ERK1 and ERK2 cascade
regulation of platelet-derived growth factor production
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-z4xvs:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0