Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDRG2 SIRNA Assays ›
Gene Symbol:
NDRG2
Gene Name:
NDRG family member 2
Gene Aliases:
SYLD
Chromosome Location:
Chr.14: 21016763 - 21070872 on Build GRCh38
UniGene ID:
Hs.525205
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS126270
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDRG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282211.1
NP_001269140.1
9
689
NM_001282212.1
NP_001269141.1
10
963
NM_001282213.1
NP_001269142.1
10
968
NM_001282214.1
NP_001269143.1
10
956
NM_001282215.1
NP_001269144.1
10
786
NM_001282216.1
NP_001269145.1
9
704
NM_001320329.1
NP_001307258.1
10
1621
NM_016250.2
NP_057334.1
10
811
NM_201535.1
NP_963293.1
11
853
NM_201536.1
NP_963294.1
9
744
NM_201537.1
NP_963831.1
10
786
NM_201538.1
NP_963832.1
9
721
NM_201539.1
NP_963833.1
10
763
NM_201540.1
NP_963834.1
11
830
NM_201541.1
NP_963835.1
10
788
XM_011536996.2
XP_011535298.1
11
840
XM_011536997.1
XP_011535299.1
11
841
XM_011536998.1
XP_011535300.1
11
831
XM_011536999.1
XP_011535301.1
9
1236
XM_011537001.1
XP_011535303.1
10
794
XM_011537002.1
XP_011535304.1
10
940
XM_017021480.1
XP_016876969.1
11
792
XM_017021481.1
XP_016876970.1
11
831
XM_017021482.1
XP_016876971.1
11
840
XM_017021483.1
XP_016876972.1
10
724
XM_017021484.1
XP_016876973.1
10
766
XM_017021485.1
XP_016876974.1
11
831
XM_017021486.1
XP_016876975.1
11
841
XM_017021487.1
XP_016876976.1
10
750
XM_017021488.1
XP_016876977.1
10
791
XM_017021489.1
XP_016876978.1
10
896
XM_017021490.1
XP_016876979.1
9
688
XM_017021491.1
XP_016876980.1
9
723
XM_017021492.1
XP_016876981.1
10
794
XM_017021493.1
XP_016876982.1
10
940
XM_017021494.1
XP_016876983.1
11
792
XM_017021495.1
XP_016876984.1
10
724
XM_017021496.1
XP_016876985.1
9
723
XM_017021497.1
XP_016876986.1
5
321
GenBank mRNA
AB033074.1
-
9
673
AF087872.1
-
9
727
AF159092.3
-
10
734
AF304051.1
-
11
853
AK024521.1
-
11
786
AK057843.1
-
10
757
AK057847.1
-
9
721
AK090657.1
-
9
660
AK096999.1
-
10
1621
AK098388.1
-
781
AK130029.1
-
9
659
AK293514.1
-
9
689
AL136574.1
-
9
698
BC010458.1
-
9
692
4215141
BC011240.1
-
10
746
4153602
BC013209.1
-
2
817
BC093038.1
-
10
787
4811718
BI759006.1
-
11
772
BM810623.1
-
10
727
BX247987.1
-
10
822
BX248031.1
-
7
563
BX647091.1
-
7
886
BX647748.1
-
10
903
CR749252.1
-
10
956
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cytokine production
signal transduction
regulation of vascular endothelial growth factor production
Wnt signaling pathway
substantia nigra development
cell differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of ERK1 and ERK2 cascade
regulation of platelet-derived growth factor production
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0