Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP28 SIRNA Assays ›
Gene Symbol:
USP28
Gene Name:
ubiquitin specific peptidase 28
Gene Aliases:
-
Chromosome Location:
Chr.11: 113797875 - 113875580 on Build GRCh38
UniGene ID:
Hs.503891
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS126600
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP28
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301029.1
NP_001287958.1
14
1746
NM_020886.3
NP_065937.1
16
1957
XM_005271630.2
XP_005271687.1
16
1956
XM_005271631.2
XP_005271688.1
16
1953
XM_005271632.2
XP_005271689.1
17
2286
XM_005271633.4
XP_005271690.1
17
2014
XM_005271636.2
XP_005271693.1
15
1776
XM_005271637.2
XP_005271694.1
16
1956
XM_005271638.2
XP_005271695.1
16
1956
XM_005271639.2
XP_005271696.1
16
1953
XM_011542936.2
XP_011541238.1
15
1875
XM_011542938.1
XP_011541240.1
14
1749
XM_011542941.1
XP_011541243.1
9
1132
XM_011542942.1
XP_011541244.1
5
672
XM_017018056.1
XP_016873545.1
15
1872
XM_017018057.1
XP_016873546.1
17
2283
XM_017018058.1
XP_016873547.1
15
1889
XM_017018059.1
XP_016873548.1
15
1872
XM_017018060.1
XP_016873549.1
14
1746
XM_017018061.1
XP_016873550.1
14
1749
XM_017018062.1
XP_016873551.1
14
1746
XM_017018063.1
XP_016873552.1
15
1833
XM_017018064.1
XP_016873553.1
14
1754
XM_017018065.1
XP_016873554.1
17
1998
XM_017018066.1
XP_016873555.1
18
2360
XM_017018067.1
XP_016873556.1
13
1609
XM_017018068.1
XP_016873557.1
9
1132
XM_017018069.1
XP_016873558.1
5
672
GenBank mRNA
AB040948.1
-
7
1100
AF266283.1
-
16
1894
AK097623.1
-
16
1587
AK304770.1
-
14
1717
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage checkpoint
DNA repair
ubiquitin-dependent protein catabolic process
cellular response to DNA damage stimulus
Ras protein signal transduction
cell proliferation
response to ionizing radiation
protein deubiquitination
regulation of protein stability
cellular response to UV
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
thiol-dependent ubiquitin-specific protease activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-n8jtk:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0