Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020964.2 | NP_066015.2 | 23 | 4271 | ||
XM_011526122.1 | XP_011524424.1 | 23 | 4271 | |||
XM_017025889.1 | XP_016881378.1 | 23 | 4253 | |||
XM_017025890.1 | XP_016881379.1 | 23 | 4253 | |||
XM_017025891.1 | XP_016881380.1 | 22 | 4130 | |||
XM_017025892.1 | XP_016881381.1 | 22 | 4296 | |||
XM_017025893.1 | XP_016881382.1 | 15 | 2143 | |||
GenBank mRNA | AB046852.1 | - | 23 | 4206 | ||
AK023817.1 | - | 7 | 950 | |||
BC130614.1 | - | 6 | 804 | |||
BC144104.1 | - | 6 | 804 | |||
BC144105.1 | - | 6 | 804 |