Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021233.2 | NP_067056.2 | 6 | 996 | ||
NM_058248.1 | NP_490649.1 | 4 | 746 | |||
XM_011541878.2 | XP_011540180.1 | 4 | 898 | |||
GenBank mRNA | AF274571.1 | - | 6 | 996 | ||
AF333389.1 | - | 4 | 746 | |||
BC065182.1 | - | 4 | 640 | |||
BC080621.1 | - | 4 | 634 | |||
BC126136.1 | - | 6 | 987 | 8991852 | ||
BC126138.1 | - | 6 | 987 | 8991854 |