Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021259.2 | NP_067082.2 | 11 | 1950 | ||
GenBank mRNA | AB045292.1 | - | 11 | 1907 | ||
AK024725.1 | - | 8 | 1249 | |||
AK091823.1 | - | 11 | 1728 | |||
BC021557.1 | - | 11 | 1841 | 4899167 |