Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRPV4 SIRNA Assays ›
          Gene Symbol:
          TRPV4
          Gene Name:
          transient receptor potential cation channel subfamily V member 4
          Gene Aliases:
          BCYM3, CMT2C, HMSN2C, OTRPC4, SMAL, SPSMA, SSQTL1, TRP12, VRL2, VROAC
          Chromosome Location:
          Chr.12: 109783087 - 109833407 on Build GRCh38
          UniGene ID:
          Hs.506713
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS126974
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRPV4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177428.1 NP_001170899.1 13 2297
          NM_001177431.1 NP_001170902.1 15 2305
          NM_001177433.1 NP_001170904.1 12 2117
          NM_021625.4 NP_067638.3 15 2496
          NM_147204.2 NP_671737.1 13 2258
          XM_005253918.1 XP_005253975.1 15 2856
          XM_011538630.2 XP_011536932.2 15 3599
          XM_011538631.2 XP_011536933.2 14 3459
          XM_011538632.2 XP_011536934.2 14 3420
          XM_011538633.2 XP_011536935.2 13 3279
          XM_017019774.1 XP_016875263.1 15 3229
          GenBank mRNA AB032427.1 - 14 2502
          AB073669.1 - 13 2227
          AB100308.1 - 14 2407
          AF258465.1 - 14 2407
          AF263523.1 - 14 2407
          AF279673.1 - 14 2203
          AJ296305.1 - 15 2496
          AL834192.1 - 15 2468
          BC117426.1 - 15 2490 40125977
          BC143307.1 - 15 2532
          BC143315.1 - 15 2490 9051820
          DQ059644.1 - 13 2266
          DQ059645.1 - 15 2305
          DQ059646.1 - 12 2086

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          diet induced thermogenesis
          calcium ion transport
          cellular calcium ion homeostasis
          cell volume homeostasis
          actin filament organization
          cell-cell junction assembly
          positive regulation of cytosolic calcium ion concentration
          osmosensory signaling pathway
          response to mechanical stimulus
          positive regulation of gene expression
          positive regulation of macrophage chemotaxis
          negative regulation of neuron projection development
          vasopressin secretion
          positive regulation of microtubule depolymerization
          actin cytoskeleton reorganization
          response to insulin
          cellular response to heat
          hyperosmotic salinity response
          glucose homeostasis
          positive regulation of vascular permeability
          cortical microtubule organization
          positive regulation of JNK cascade
          microtubule polymerization
          regulation of response to osmotic stress
          positive regulation of inflammatory response
          multicellular organismal water homeostasis
          positive regulation of ERK1 and ERK2 cascade
          calcium ion import
          calcium ion transmembrane transport
          cellular response to osmotic stress
          cellular hypotonic response
          cellular hypotonic salinity response
          positive regulation of monocyte chemotactic protein-1 production
          positive regulation of macrophage inflammatory protein 1 alpha production
          positive regulation of chemokine (C-C motif) ligand 5 production
          blood vessel endothelial cell delamination
          negative regulation of brown fat cell differentiation
          signal transduction involved in regulation of aerobic respiration
          positive regulation of chemokine (C-X-C motif) ligand 1 production
          positive regulation of energy homeostasis
          positive regulation of interleukin-6 secretion
          actin binding
          osmosensor activity
          protein kinase C binding
          cation channel activity
          calcium channel activity
          protein binding
          calmodulin binding
          ATP binding
          microtubule binding
          stretch-activated, cation-selective, calcium channel activity
          protein kinase binding
          SH2 domain binding
          alpha-tubulin binding
          beta-tubulin binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0