Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RXFP1 SIRNA Assays ›
Gene Symbol:
RXFP1
Gene Name:
relaxin/insulin like family peptide receptor 1
Gene Aliases:
LGR7, RXFPR1
Chromosome Location:
Chr.4: 158521711 - 158653372 on Build GRCh38
UniGene ID:
Hs.591686
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS126999
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RXFP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001253727.1
NP_001240656.1
12, 13
1305
NM_001253728.1
NP_001240657.1
11, 12
1125
NM_001253730.1
NP_001240659.1
11, 12
1149
NM_001253732.1
NP_001240661.1
13, 14
1308
NM_001253733.1
NP_001240662.1
12, 13
1236
NM_021634.3
NP_067647.2
12, 13
1224
NR_045579.1
12, 13
1841
NR_045580.1
12, 13
1277
NR_045581.1
12, 13
1248
NR_045583.1
11, 12
1167
NR_045584.1
13, 14
1277
XM_011532174.1
XP_011530476.1
12, 13
1274
XM_011532175.1
XP_011530477.1
11, 12
1202
XM_011532176.1
XP_011530478.1
11, 12
1124
XM_011532179.2
XP_011530481.1
12, 13
1274
XM_017008517.1
XP_016864006.1
11, 12
1202
XM_017008518.1
XP_016864007.1
12, 13
1196
XM_017008519.1
XP_016864008.1
13, 14
1238
XM_017008520.1
XP_016864009.1
12, 13
1327
XM_017008521.1
XP_016864010.1
13, 14
1157
XM_017008522.1
XP_016864011.1
12, 13
1245
XM_017008523.1
XP_016864012.1
12, 13
1196
XM_017008524.1
XP_016864013.1
11, 12
1124
XM_017008525.1
XP_016864014.1
11, 12
1128
XM_017008526.1
XP_016864015.1
7, 8
695
GenBank mRNA
AB208943.1
-
13, 14
1046
AF190500.1
-
12, 13
961
AK122647.1
-
14, 15
1265
AK294694.1
-
12, 13
1069
AK295040.1
-
12, 13
1664
AK300379.1
-
11, 12
1020
AK307889.1
-
11, 12
1066
AK312470.1
-
12, 13
1068
AY899848.1
-
13, 14
1014
AY899850.1
-
11, 12
862
BC112142.1
-
12, 13
1043
8327610
BC113617.1
-
12, 13
1046
8322669
BC143556.1
-
12, 13
1066
BC143558.1
-
12, 13
1099
BC143559.1
-
11, 12
971
BX647975.1
-
12, 13
1162
BX647980.1
-
12, 13
1162
BX647985.1
-
12, 13
1191
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0