Search
Search

TMEM35A| Gene Symbol: TMEM35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021637.2 | NP_067650.1 | 2 | 338 | ||
| GenBank mRNA | AK024146.1 | - | 2 | 311 | ||
| BC050273.1 | - | 2 | 303 | 4801325 | ||
| BC078658.1 | - | 2 | 303 | 5261160 | ||