Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321881.1 | NP_001308810.1 | 11 | 948 | ||
NM_001321882.1 | NP_001308811.1 | 11 | 1166 | |||
NM_024635.3 | NP_078911.3 | 11 | 952 | |||
XM_005252127.3 | XP_005252184.1 | 11 | 1023 | |||
XM_011518903.2 | XP_011517205.1 | 8 | 755 | |||
XM_011518904.2 | XP_011517206.1 | 5 | 441 | |||
XM_017015012.1 | XP_016870501.1 | 12 | 1011 | |||
GenBank mRNA | AK025266.1 | - | 11 | 948 | ||
AK056059.1 | - | 11 | 1166 | |||
AK225237.1 | - | 11 | 948 | |||
AK292239.1 | - | 11 | 952 | |||
BC117427.1 | - | 11 | 873 | 40125978 | ||
BC117429.1 | - | 11 | 869 | 40125980 |