Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172673.1 | NP_001166144.1 | 4, 5 | 561 | ||
NM_021938.3 | NP_068757.2 | 4, 5 | 562 | |||
NR_033342.1 | 4, 5 | 400 | ||||
XM_006722832.1 | XP_006722895.1 | 4, 5 | 589 | |||
XM_006722833.1 | XP_006722896.1 | 4, 5 | 586 | |||
XM_006722836.2 | XP_006722899.1 | 3, 4 | 230 | |||
XM_006722837.3 | XP_006722900.1 | 4, 5 | 501 | |||
XM_006722838.3 | XP_006722901.1 | 3, 4 | 373 | |||
XM_011528173.1 | XP_011526475.1 | 3, 4 | 202 | |||
XM_011528174.2 | XP_011526476.1 | 3, 4 | 371 | |||
XM_017027111.1 | XP_016882600.1 | 2, 3 | 445 | |||
GenBank mRNA | AB209067.1 | - | 4, 5 | 561 | ||
AF329266.1 | - | 4, 5 | 525 | |||
AK294109.1 | - | 4, 5 | 400 | |||
AL832550.1 | - | 2, 3 | 150 | |||
BC028101.1 | - | 4, 5 | 562 | 5242909 |