Search
Search

GALNT11| Gene Symbol: GALNT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304514.1 | NP_001291443.1 | 6 | 947 | ||
| NM_022087.3 | NP_071370.2 | 7 | 1280 | |||
| XM_006716082.2 | XP_006716145.1 | 7 | 1191 | |||
| XM_006716083.2 | XP_006716146.1 | 7 | 1159 | |||
| XM_006716084.2 | XP_006716147.1 | 7 | 1123 | |||
| XM_017012499.1 | XP_016867988.1 | 7 | 1261 | |||
| XM_017012500.1 | XP_016867989.1 | 6 | 1137 | |||
| GenBank mRNA | AK025287.1 | - | 7 | 1114 | ||
| AK026056.1 | - | 3 | 353 | |||
| AK124934.1 | - | 9 | 1468 | |||
| AK128545.1 | - | 2 | 1106 | |||
| AK298911.1 | - | 6 | 870 | |||
| Y12434.3 | - | 6 | 1031 | |||