Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB41L4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022140.3 | NP_071423.3 | 12 | 1298 | ||
XM_011543530.1 | XP_011541832.1 | 12 | 1475 | |||
XM_011543531.2 | XP_011541833.1 | 12 | 1475 | |||
XM_011543532.1 | XP_011541834.1 | 12 | 1475 | |||
XM_011543533.1 | XP_011541835.1 | 12 | 1475 | |||
XM_017009689.1 | XP_016865178.1 | 12 | 1475 | |||
GenBank mRNA | AA350575.1 | - | 2 | 82 | ||
AB030240.1 | - | 12 | 1027 | |||
BC114632.1 | - | 12 | 1124 | |||
BC114942.1 | - | 12 | 1119 |