Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGAP9 SIRNA Assays ›
Gene Symbol:
ARHGAP9
Gene Name:
Rho GTPase activating protein 9
Gene Aliases:
10C, RGL1
Chromosome Location:
Chr.12: 57472255 - 57488814 on Build GRCh38
UniGene ID:
Hs.437126
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS127538
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGAP9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080156.2
NP_001073625.1
14
1690
NM_001080157.1
NP_001073626.1
15
2045
NM_001319850.1
NP_001306779.1
19
2273
NM_001319851.1
NP_001306780.1
13
1966
NM_001319852.1
NP_001306781.1
14
1747
NM_032496.3
NP_115885.2
16
1971
XM_005269083.2
XP_005269140.1
14
1412
XM_005269084.2
XP_005269141.1
13
1345
XM_005269085.2
XP_005269142.1
13
1319
XM_011538656.2
XP_011536958.1
18
2008
XM_011538657.2
XP_011536959.1
15
1849
XM_011538658.2
XP_011536960.1
15
1791
XM_011538659.2
XP_011536961.1
18
2008
XM_017019800.1
XP_016875289.1
15
2285
XM_017019801.1
XP_016875290.1
14
1355
XM_017019802.1
XP_016875291.1
13
1288
XM_017019803.1
XP_016875292.1
14
1355
XM_017019804.1
XP_016875293.1
13
1288
GenBank mRNA
AB030239.1
-
14
1442
AB051853.1
-
16
1949
AK057573.1
-
13
1619
AK092763.1
-
15
2036
AK131415.1
-
19
2200
AK301095.1
-
14
1342
AL713803.1
-
9
1034
BC006107.1
-
19
2273
3010091
BC011820.1
-
14
1416
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity
protein binding
phosphatidylinositol-3,4,5-trisphosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0