Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127895.1 | NP_001121367.1 | 3 | 550 | ||
NM_001127896.1 | NP_001121368.1 | 2 | 461 | |||
NM_022467.3 | NP_071912.2 | 2 | 803 | |||
XM_011527222.1 | XP_011525524.1 | 2 | 784 | |||
XM_011527224.1 | XP_011525526.1 | 3 | 918 | |||
XM_017027143.1 | XP_016882632.1 | 2 | 454 | |||
GenBank mRNA | AB047801.1 | - | 2 | 317 | ||
AF300612.1 | - | 2 | 264 | |||
AF305781.1 | - | 2 | 360 | |||
BC011380.1 | - | 2 | 413 | 3874578 | ||
BC014250.2 | - | 2 | 457 | 4635405 | ||
BC018723.1 | - | 2 | 803 | 4137487 | ||
BI917117.1 | - | 2 | 461 | |||
BX354555.2 | - | 3 | 549 | |||
DB500322.1 | - | 2 | 473 |