Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DCLRE1C SIRNA Assays ›
Gene Symbol:
DCLRE1C
Gene Name:
DNA cross-link repair 1C
Gene Aliases:
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C
Chromosome Location:
Chr.10: 14897853 - 14954432 on Build GRCh38
UniGene ID:
Hs.655932
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS127606
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DCLRE1C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033855.2
NP_001029027.1
6, 7
896
NM_001033857.2
NP_001029029.1
7, 8
981
NM_001033858.2
NP_001029030.1
8, 9
1303
NM_001289076.1
NP_001276005.1
4, 5
755
NM_001289077.1
NP_001276006.1
7, 8
1027
NM_001289078.1
NP_001276007.1
4, 5
788
NM_001289079.1
NP_001276008.1
8, 9
1349
NM_022487.3
NP_071932.2
5, 6
840
NR_110297.1
8, 9
1108
XM_006717491.3
XP_006717554.1
6, 7
570
XM_011519616.1
XP_011517918.1
6, 7
586
XM_011519617.1
XP_011517919.1
7, 8
705
XM_011519619.1
XP_011517921.1
8, 9
732
XM_011519620.2
XP_011517922.1
6, 7
571
XM_011519621.1
XP_011517923.1
6, 7
571
XM_017016556.1
XP_016872045.1
6, 7
571
XM_017016557.1
XP_016872046.1
6, 7
598
XM_017016558.1
XP_016872047.1
7, 8
644
XM_017016559.1
XP_016872048.1
4, 5
439
XM_017016560.1
XP_016872049.1
7, 8
660
GenBank mRNA
AF395747.1
-
7, 8
597
AF395748.1
-
7, 8
643
AF395749.1
-
5, 6
456
AF395750.1
-
4, 5
404
AF395751.1
-
8, 9
919
AF395752.1
-
8, 9
965
AJ296101.1
-
6, 7
512
AK021422.1
-
5, 6
503
AK022922.1
-
8, 9
902
AK290598.1
-
6, 7
559
AK307912.1
-
6, 7
558
AW954867.1
-
5, 6
354
BC009185.2
-
4, 5
379
3945623
BC022254.1
-
6, 7
496
4272155
BE799340.1
-
4, 5
402
BX648858.1
-
7, 8
613
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
double-strand break repair via nonhomologous end joining
response to ionizing radiation
B cell differentiation
protection from non-homologous end joining at telomere
V(D)J recombination
interstrand cross-link repair
nucleic acid phosphodiester bond hydrolysis
single-stranded DNA endodeoxyribonuclease activity
damaged DNA binding
endonuclease activity
protein binding
5'-3' exonuclease activity
5'-3' exodeoxyribonuclease activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0