Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198978.1 | NP_001185907.1 | 9 | 1118 | ||
NM_001198979.1 | NP_001185908.1 | 9 | 1202 | |||
NM_001198980.1 | NP_001185909.1 | 9 | 1133 | |||
NM_022733.2 | NP_073570.1 | 9 | 1560 | |||
XM_011541960.1 | XP_011540262.1 | 10 | 1567 | |||
XM_017002093.1 | XP_016857582.1 | 6 | 1259 | |||
GenBank mRNA | AK294675.1 | - | 4 | 842 | ||
AK298702.1 | - | 9 | 1098 | |||
AK303975.1 | - | 9 | 1106 | |||
AK308280.1 | - | 9 | 1203 | |||
AK313105.1 | - | 9 | 1507 | |||
AK316218.1 | - | 9 | 1133 | |||
AL133206.1 | - | 9 | 1074 | |||
AL137764.1 | - | 9 | 1087 | |||
BC014512.1 | - | 7 | 899 | |||
BC021133.1 | - | 9 | 1210 | 4855320 |