Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011554.2 | NP_001011554.1 | 14 | 1818 | ||
NM_001193339.1 | NP_001180268.1 | 12 | 1632 | |||
NM_001193340.1 | NP_001180269.1 | 13 | 1713 | |||
NM_001193342.1 | NP_001180271.1 | 13 | 1609 | |||
NM_022829.5 | NP_073740.2 | 13 | 1782 | |||
GenBank mRNA | AF154121.1 | - | 13 | 1761 | ||
AK056713.1 | - | 13 | 1780 | |||
AK096658.1 | - | 13 | 1713 | |||
AK123351.1 | - | 2 | 406 | |||
AK293902.1 | - | 13 | 1598 | |||
AK295748.1 | - | 12 | 1630 | |||
AK303709.1 | - | 4 | 504 | |||
AK310767.1 | - | 3 | 694 | |||
AK310960.1 | - | 12 | 1791 | |||
AK315131.1 | - | 13 | 1780 | |||
AL442082.1 | - | 4 | 446 | |||
AY072810.3 | - | 13 | 1761 | |||
BC035966.1 | - | 13 | 1780 | 4609776 |