Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022897.4 | NP_075048.1 | 20 | 2338 | ||
XM_011534627.1 | XP_011532929.1 | 20 | 2194 | |||
XM_011534631.2 | XP_011532933.1 | 19 | 2207 | |||
XM_017009736.1 | XP_016865225.1 | 19 | 2074 | |||
XM_017009737.1 | XP_016865226.1 | 19 | 2207 | |||
XM_017009738.1 | XP_016865227.1 | 19 | 2047 | |||
XM_017009739.1 | XP_016865228.1 | 19 | 2047 | |||
XM_017009740.1 | XP_016865229.1 | 18 | 1927 | |||
XM_017009741.1 | XP_016865230.1 | 20 | 2194 | |||
XM_017009742.1 | XP_016865231.1 | 20 | 2194 | |||
XM_017009743.1 | XP_016865232.1 | 20 | 2368 | |||
XM_017009744.1 | XP_016865233.1 | 20 | 2194 | |||
XM_017009745.1 | XP_016865234.1 | 12 | 1391 | |||
XM_017009747.1 | XP_016865236.1 | 8 | 666 | |||
GenBank mRNA | AF222747.1 | - | 20 | 2194 | ||
AJ271459.1 | - | 20 | 2202 | |||
AJ288952.1 | - | 23 | 2541 | |||
AJ288953.1 | - | 22 | 2380 | |||
AJ288954.1 | - | 22 | 2501 | |||
AJ288955.1 | - | 21 | 2320 | |||
AK027880.1 | - | 18 | 2022 | |||
AK057478.1 | - | 5 | 533 | |||
AK315159.1 | - | 21 | 2406 | |||
AL832980.1 | - | 20 | 2237 |