Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022900.4 | NP_075051.4 | 8 | 921 | ||
XM_006716092.3 | XP_006716155.1 | 6 | 416 | |||
XM_006716093.3 | XP_006716156.2 | 8 | 759 | |||
XM_011516495.2 | XP_011514797.1 | 8 | 759 | |||
GenBank mRNA | AF355594.1 | - | 8 | 714 | ||
AF397424.2 | - | 8 | 841 | |||
AK025532.1 | - | 1 | 12 | |||
AK123866.1 | - | 8 | 759 | |||
AK314329.1 | - | 8 | 719 | |||
BC063284.1 | - | 8 | 906 | 5286382 |