Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNK2 SIRNA Assays ›
Gene Symbol:
WNK2
Gene Name:
WNK lysine deficient protein kinase 2
Gene Aliases:
NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43
Chromosome Location:
Chr.9: 93184156 - 93327581 on Build GRCh38
UniGene ID:
Hs.654856
Species:
Human
siRNA ID
HSS128038
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WNK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282394.1
NP_001269323.1
4
1125
NM_006648.3
NP_006639.3
4
1125
XM_005252137.3
XP_005252194.1
5
1357
XM_005252140.2
XP_005252197.1
5
1357
XM_005252141.2
XP_005252198.1
5
1357
XM_005252144.2
XP_005252201.1
5
1357
XM_011518926.2
XP_011517228.1
5
1357
XM_011518927.2
XP_011517229.1
5
1357
XM_011518928.2
XP_011517230.1
5
1357
XM_011518929.2
XP_011517231.1
5
1357
XM_011518930.2
XP_011517232.1
5
1357
XM_011518931.2
XP_011517233.1
5
1357
XM_011518932.2
XP_011517234.1
5
1357
XM_011518935.2
XP_011517237.1
5
1357
XM_011518936.2
XP_011517238.1
5
1357
XM_011518937.2
XP_011517239.1
4
665
XM_011518939.2
XP_011517241.1
4
710
XM_011518940.2
XP_011517242.1
4
712
XM_017015045.1
XP_016870534.1
5
1357
XM_017015046.1
XP_016870535.1
5
1357
XM_017015047.1
XP_016870536.1
5
1357
XM_017015048.1
XP_016870537.1
5
1357
XM_017015049.1
XP_016870538.1
5
1357
XM_017015050.1
XP_016870539.1
5
1357
XM_017015051.1
XP_016870540.1
5
1357
XM_017015052.1
XP_016870541.1
5
1357
XM_017015053.1
XP_016870542.1
5
1357
XM_017015054.1
XP_016870543.1
5
1357
XM_017015055.1
XP_016870544.1
5
1357
GenBank mRNA
AB044546.1
-
4
1149
AB051547.2
-
4
1112
AJ242724.4
-
4
1125
AK310390.1
-
5
550
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
negative regulation of cell proliferation
intracellular signal transduction
protein autophosphorylation
negative regulation of ERK1 and ERK2 cascade
regulation of ion homeostasis
positive regulation of sodium ion transmembrane transporter activity
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-zbtnl:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0