Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAPRIN2 SIRNA Assays ›
Gene Symbol:
CAPRIN2
Gene Name:
caprin family member 2
Gene Aliases:
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location:
Chr.12: 30709552 - 30754994 on Build GRCh38
UniGene ID:
Hs.234355
Species:
Human
siRNA ID
HSS128053
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAPRIN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002259.2
NP_001002259.1
18
3674
NM_001206856.2
NP_001193785.1
17
3550
NM_001319842.1
NP_001306771.1
17
3588
NM_001319843.1
NP_001306772.1
18
3671
NM_001319844.1
NP_001306773.1
18
2824
NM_001319845.1
NP_001306774.1
19
2929
NM_001319846.1
NP_001306775.1
19
2919
NM_023925.4
NP_076414.2
17
3524
NM_032156.4
NP_115532.3
18
3715
XM_006719138.3
XP_006719201.1
18
3734
XM_006719139.3
XP_006719202.1
17
3692
XM_006719140.3
XP_006719203.1
17
3689
XM_006719142.3
XP_006719205.1
17
3587
XM_006719144.3
XP_006719207.1
16
3545
XM_006719145.3
XP_006719208.1
18
3841
XM_006719146.3
XP_006719209.1
18
3873
XM_006719147.3
XP_006719210.1
18
3778
XM_017019851.1
XP_016875340.1
17
3629
XM_017019852.1
XP_016875341.1
19
2889
XM_017019853.1
XP_016875342.1
19
2959
XM_017019854.1
XP_016875343.1
16
3542
XM_017019855.1
XP_016875344.1
19
2856
XM_017019856.1
XP_016875345.1
18
2814
XM_017019857.1
XP_016875346.1
18
2784
XM_017019858.1
XP_016875347.1
18
2811
XM_017019859.1
XP_016875348.1
17
2640
XM_017019860.1
XP_016875349.1
19
3841
XM_017019861.1
XP_016875350.1
17
2664
XM_017019862.1
XP_016875351.1
19
3838
XM_017019863.1
XP_016875352.1
19
3885
XM_017019865.1
XP_016875354.1
17
3736
XM_017019866.1
XP_016875355.1
19
2936
XM_017019867.1
XP_016875356.1
19
2963
XM_017019868.1
XP_016875357.1
19
2933
XM_017019869.1
XP_016875358.1
19
2960
XM_017019870.1
XP_016875359.1
19
2873
XM_017019871.1
XP_016875360.1
19
2944
XM_017019872.1
XP_016875361.1
18
2795
XM_017019873.1
XP_016875362.1
17
2711
XM_017019874.1
XP_016875363.1
18
2806
XM_017019875.1
XP_016875364.1
17
2648
XM_017019876.1
XP_016875365.1
16
2357
XM_017019877.1
XP_016875366.1
15
2315
XM_017019878.1
XP_016875367.1
15
2312
XM_017019879.1
XP_016875368.1
17
2641
XM_017019880.1
XP_016875369.1
14
2105
XM_017019881.1
XP_016875370.1
18
2850
XM_017019882.1
XP_016875371.1
18
2790
XM_017019883.1
XP_016875372.1
15
2359
GenBank mRNA
AB058776.1
-
3161
AF525416.1
-
10
1185
AK024247.1
-
5
2641
AK026222.1
-
4
490
AL834212.1
-
16
2202
AY074490.1
-
17
3524
AY074491.1
-
17
3588
BC007520.1
-
4
395
BC066295.1
-
4
426
BC111007.1
-
4
478
BC117672.1
-
18
3512
40068103
BC117673.1
-
17
3347
40068104
BX537569.1
-
15
5730
BX538080.1
-
5
610
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of translation
negative regulation of cell growth
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of dendrite morphogenesis
positive regulation of dendritic spine morphogenesis
positive regulation of canonical Wnt signaling pathway
RNA binding
receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-5svh6:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0