Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC25A23 SIRNA Assays ›
Gene Symbol:
SLC25A23
Gene Name:
solute carrier family 25 member 23
Gene Aliases:
APC2, MCSC2, SCaMC-3
Chromosome Location:
Chr.19: 6436081 - 6460221 on Build GRCh38
UniGene ID:
Hs.356231
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS128238
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC25A23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024103.2
NP_077008.2
6
872
XM_011528274.1
XP_011526576.1
7
1464
XM_011528275.1
XP_011526577.1
6
1323
XM_011528276.1
XP_011526578.1
7
1464
XM_011528277.1
XP_011526579.1
7
1464
XM_011528278.1
XP_011526580.1
7
1464
XM_011528279.1
XP_011526581.1
7
1464
XM_011528280.1
XP_011526582.1
7
1464
XM_011528281.1
XP_011526583.1
7
1464
XM_011528282.1
XP_011526584.1
7
1464
XM_011528283.1
XP_011526585.1
7
1464
XM_011528284.1
XP_011526586.1
7
1464
XM_011528285.1
XP_011526587.1
7
1464
XM_017027285.1
XP_016882774.1
7
1464
XM_017027286.1
XP_016882775.1
6
1323
XM_017027287.1
XP_016882776.1
6
1323
XM_017027288.1
XP_016882777.1
6
1323
XM_017027289.1
XP_016882778.1
7
1464
XM_017027290.1
XP_016882779.1
6
1323
XM_017027291.1
XP_016882780.1
6
1323
XM_017027292.1
XP_016882781.1
6
1323
GenBank mRNA
AJ512835.1
-
6
837
AJ619962.1
-
6
827
AJ619988.1
-
6
837
AK054901.1
-
5
569
AK294514.1
-
6
730
AY750170.1
-
6
837
BC001656.1
-
7
994
3356718
Back To Top
More Information
Panther Classification:
Molecular Function -
mitochondrial carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of oxidative phosphorylation
translation
mitochondrial calcium ion transport
calcium ion transmembrane import into mitochondrion
regulation of cellular respiration
regulation of sequestering of calcium ion
adenine nucleotide transport
cellular response to calcium ion
urea homeostasis
structural constituent of ribosome
calcium ion binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0