Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146056.2 | NP_001139528.1 | 6 | 1539 | ||
NM_001146083.2 | NP_001139555.1 | 7 | 1618 | |||
NM_024298.4 | NP_077274.3 | 8 | 1745 | |||
XM_011527299.2 | XP_011525601.1 | 8 | 1949 | |||
XM_011527300.2 | XP_011525602.1 | 8 | 1340 | |||
GenBank mRNA | AF211969.1 | - | 2 | 199 | ||
AK055908.1 | - | 8 | 1678 | |||
AK122789.1 | - | 9 | 1463 | |||
AK123290.1 | - | 2 | 2192 | |||
AK293199.1 | - | 6 | 1149 | |||
AK298689.1 | - | 6 | 1225 | |||
AK311493.1 | - | 7 | 1021 | |||
BC002512.2 | - | 7 | 1195 | |||
BC003164.1 | - | 8 | 1276 | 2958347 | ||
BC015857.1 | - | 7 | 1182 | 4671596 | ||
EU016381.1 | - | 7 | 1197 | |||
S82470.1 | - | 6 | 1051 |