Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024333.2 | NP_077309.1 | 11, 12 | 1450 | ||
GenBank mRNA | AF316829.1 | - | 11, 12 | 1341 | ||
AK021750.1 | - | 11, 12 | 1350 | |||
AK315661.1 | - | 11, 12 | 1444 | |||
AY032617.1 | - | 11, 12 | 1373 | |||
BC003124.1 | - | 11, 12 | 1350 | 3502614 | ||
BC016442.1 | - | 5, 6 | 1054 |