Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167911.1 | NP_001161383.1 | 3 | 492 | ||
NM_001167912.1 | NP_001161384.1 | 3 | 492 | |||
NM_001167915.1 | NP_001161387.1 | 4 | 839 | |||
NM_001167916.1 | NP_001161388.1 | 3 | 577 | |||
NM_001167917.1 | NP_001161389.1 | 2 | 421 | |||
NM_024621.2 | NP_078897.2 | 3 | 683 | |||
XM_011513134.2 | XP_011511436.1 | 2 | 426 | |||
XM_011513135.1 | XP_011511437.1 | 3 | 322 | |||
GenBank mRNA | AB051479.1 | - | 3 | 492 | ||
AK027625.1 | - | 3 | 581 | |||
AL713656.1 | - | 2 | 280 | |||
BC057999.1 | - | 3 | 580 | |||
BC101660.1 | - | 2 | 319 | 8069166 | ||
BC111017.1 | - | 3 | 484 | 5174889 | ||
BC113555.1 | - | 2 | 319 | 8322607 | ||
BG194590.1 | - | 1 | 118 | |||
DA887051.1 | - | 3 | 514 |