Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024654.4 | NP_078930.3 | 4 | 884 | ||
GenBank mRNA | AK091284.1 | - | 4 | 821 | ||
AK289653.1 | - | 4 | 816 | |||
BC094836.1 | - | 3 | 259 | |||
BC105095.1 | - | 4 | 884 | 8144098 | ||
BC110849.1 | - | 4 | 793 | 6167575 | ||
BC112278.1 | - | 4 | 884 | 8327746 | ||
BC143646.1 | - | 4 | 884 |